Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lck | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001162432.1 | NP_001155904.1 | 8 - 9 | 868 | 63 | |
NM_001162433.1 | NP_001155905.1 | 8 - 9 | 910 | 63 | ||
NM_010693.3 | NP_034823.1 | 7 - 8 | 965 | 63 | ||
GenBank mRNA | AK088001.1 | - | 7 - 8 | 966 | 63 | |
AK166263.1 | - | 2 - 3 | 355 | 63 | ||
BC011474.1 | - | 8 - 9 | 875 | 4191610 | 63 | |
M12056.1 | - | 7 - 8 | 941 | 63 | ||
X03533.1 | - | 7 - 8 | 971 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |