Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400011620.mm.1 |
NM_029219.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400011620.mm.1 |
NM_029219.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400011620.mm.1 |
TC0400001510.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400011617.mm.1 |
NM_029219.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400011617.mm.1 |
NM_029219.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400011617.mm.1 |
TC0400001510.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400011618.mm.1 |
NM_029219.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400011618.mm.1 |
NM_029219.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400011618.mm.1 |
TC0400001510.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400011619.mm.1 |
NM_029219.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400011619.mm.1 |
NM_029219.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400011619.mm.1 |
TC0400001510.mm.1 |
MTA TC |