Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plagl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009538.2 | NP_033564.2 | 2 - 3 | 279 | 134 | |
GenBank mRNA | AK142052.1 | - | 3 - 4 | 271 | 134 | |
AK142210.1 | - | 2 - 3 | 237 | 205 | ||
BC065150.1 | - | 2 - 3 | 279 | 134 | ||
BC141284.1 | - | 2 - 3 | 72 | 9055896 | 183 | |
BC145612.1 | - | 2 - 3 | 72 | 9054178 | 183 | |
X95503.1 | - | 3 - 4 | 195 | 197 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |