Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rgs17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161822.1 | NP_001155294.1 | 2 - 3 | 243 | 124 | |
GenBank mRNA | AK045770.1 | - | 2 - 3 | 210 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |