Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198914.1 | NP_001185843.1 | 5 - 6 | 788 | 65 | |
NM_010848.3 | NP_034978.3 | 5 - 6 | 788 | 65 | ||
GenBank mRNA | AK088079.1 | - | 5 - 6 | 709 | 65 | |
AK088598.1 | - | 5 - 6 | 710 | 65 | ||
AK132250.1 | - | 5 - 6 | 653 | 65 | ||
AK137965.1 | - | 5 - 6 | 654 | 65 | ||
BC011513.1 | - | 5 - 6 | 696 | 3672769 | 65 | |
M12848.1 | - | 5 - 6 | 786 | 65 | ||
X02774.1 | - | 5 - 6 | 558 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis Mammalian Gene Collection Mus musculus Probe spans exons |