Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srsf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020587.2 | NP_065612.2 | 1 - 2 | 229 | 77 | |
GenBank mRNA | AK075769.1 | - | 1 - 2 | 197 | 77 | |
AK077648.1 | - | 1 - 2 | 215 | 77 | ||
AK156989.1 | - | 1 - 2 | 164 | 77 | ||
BC019437.1 | - | 1 - 2 | 189 | 4018178 | 77 | |
BY107258.1 | - | 1 - 2 | 230 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |