Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mknk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285487.1 | NP_001272416.1 | 9 - 10 | 839 | 72 | |
NM_001285488.1 | NP_001272417.1 | 8 - 9 | 733 | 72 | ||
NM_021461.5 | NP_067436.1 | 7 - 8 | 689 | 72 | ||
GenBank mRNA | AK010697.1 | - | 7 - 8 | 667 | 72 | |
AK141298.1 | - | 7 - 8 | 667 | 72 | ||
AK155934.1 | - | 9 - 10 | 800 | 72 | ||
AK169982.1 | - | 8 - 9 | 699 | 72 | ||
BC021369.1 | - | 7 - 8 | 645 | 5030044 | 72 | |
BQ551007.1 | - | 5 - 6 | 396 | 72 | ||
Y11091.1 | - | 7 - 8 | 654 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |