Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sepn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029100.2 | NP_083376.2 | 10 - 11 | 1441 | 61 | |
GenBank mRNA | BC043665.1 | - | 2 - 3 | 129 | 61 | |
BU057838.1 | - | 2 - 3 | 128 | 61 | ||
CF737950.1 | - | 4 - 5 | 483 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |