Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map7d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145970.1 | NP_001139442.1 | 10 - 11 | 1949 | 88 | |
NM_144941.3 | NP_659190.3 | 10 - 11 | 2165 | 88 | ||
GenBank mRNA | AK122462.1 | - | 4 - 5 | 3588 | 88 | |
AK151613.1 | - | 8 - 9 | 1680 | 88 | ||
BC016081.1 | - | 1 - 2 | 201 | 88 | ||
BC019977.1 | - | 5 - 6 | 1141 | 4187506 | 88 | |
BC023677.1 | - | 10 - 11 | 2140 | 5354497 | 88 | |
BC023774.1 | - | 10 - 11 | 1946 | 88 | ||
BM944422.1 | - | 1 - 2 | 55 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |