Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hyi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_003946.2 | 4 - 5 | 474 | 93 | ||
GenBank mRNA | AK004328.1 | - | 3 - 4 | 174 | 93 | |
AK020099.1 | - | 4 - 5 | 460 | 93 | ||
CB952524.1 | - | 2 - 3 | 91 | 93 | ||
CK620463.1 | - | 4 - 5 | 474 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Mus musculus Probe spans exons |