Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcf7l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079822.2 | NP_001073290.1 | 6 - 7 | 790 | 78 | |
NM_009332.3 | NP_033358.2 | 5 - 6 | 748 | 78 | ||
GenBank mRNA | AF057691.1 | - | 1 - 2 | 136 | 78 | |
AJ223069.1 | - | 5 - 6 | 656 | 78 | ||
AK052721.1 | - | 5 - 6 | 958 | 78 | ||
BC128305.1 | - | 6 - 7 | 697 | 40109611 | 78 | |
BC128306.1 | - | 5 - 6 | 655 | 40109613 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons |