Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clec1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001204253.1 | NP_001191182.1 | 2 - 3 | 510 | 101 | |
GenBank mRNA | EF070192.1 | - | 2 - 3 | 197 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |