Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400027133.mm.1 |
TC0400004218.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400027133.mm.1 |
NM_001034876.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400027133.mm.1 |
NM_001034876.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400027129.mm.1 |
TC0400004218.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400027129.mm.1 |
NM_001034876.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400027129.mm.1 |
NM_001034876.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400027128.mm.1 |
TC0400004218.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400027128.mm.1 |
NM_001034876.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400027128.mm.1 |
NM_001034876.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400027130.mm.1 |
TC0400004218.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400027130.mm.1 |
NM_001034876.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400027130.mm.1 |
NM_001034876.1 |
Refseq |