Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nagk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164187.1 | NP_001157659.1 | 1 - 2 | 218 | 83 | |
GenBank mRNA | AJ242911.1 | - | 1 - 2 | 179 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mus musculus Probe spans exons |