Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sult3a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020565.2 | NP_065590.2 | 3 - 4 | 411 | 172 | |
GenBank mRNA | AF026075.1 | - | 3 - 4 | 412 | 172 | |
BC125593.1 | - | 3 - 4 | 411 | 40130108 | 172 | |
BC125595.1 | - | 3 - 4 | 411 | 40130110 | 172 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |