Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zufsp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028287.2 | NP_082563.1 | 6 - 7 | 1173 | 110 | |
GenBank mRNA | AK046191.1 | - | 6 - 7 | 1176 | 110 | |
AK052071.1 | - | 6 - 7 | 1157 | 110 | ||
AK077304.1 | - | 6 - 7 | 1221 | 110 | ||
AK077730.1 | - | 6 - 7 | 1194 | 110 | ||
AK160928.1 | - | 6 - 7 | 1172 | 110 | ||
BC089514.1 | - | 6 - 7 | 1186 | 30252639 | 110 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |