Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nsg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290680.1 | NP_001277609.1 | 3 - 4 | 275 | 57 | |
NM_001290681.1 | NP_001277610.1 | 3 - 4 | 271 | 57 | ||
NM_008741.4 | NP_032767.1 | 3 - 4 | 411 | 57 | ||
GenBank mRNA | AK011563.1 | - | 2 - 3 | 227 | 57 | |
AK075612.1 | - | 3 - 4 | 368 | 57 | ||
AK088499.1 | - | 3 - 4 | 403 | 57 | ||
BC018224.1 | - | 3 - 4 | 343 | 4527900 | 57 | |
BU058771.1 | - | 3 - 4 | 274 | 57 | ||
DN176718.1 | - | 3 - 4 | 411 | 57 | ||
U17259.1 | - | 3 - 4 | 371 | 57 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |