Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Thnsl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178413.5 | NP_848500.3 | 1 - 2 | 91 | 68 | |
GenBank mRNA | BC025604.1 | - | 1 - 2 | 54 | 5250319 | 68 |
CB600010.1 | - | 1 - 2 | 61 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |