Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0600023454.mm.1 |
TC0600003038.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600023454.mm.1 |
NM_198884.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600023454.mm.1 |
NM_198884.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600023450.mm.1 |
TC0600003038.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600023450.mm.1 |
NM_198884.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600023450.mm.1 |
NM_198884.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600023451.mm.1 |
TC0600003038.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600023451.mm.1 |
NM_198884.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600023451.mm.1 |
NM_198884.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600023452.mm.1 |
TC0600003038.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600023452.mm.1 |
NM_198884.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600023452.mm.1 |
NM_198884.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600023455.mm.1 |
TC0600003038.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600023455.mm.1 |
NM_198884.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600023455.mm.1 |
NM_198884.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600023457.mm.1 |
TC0600003038.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600023457.mm.1 |
NM_198884.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600023457.mm.1 |
NM_198884.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600023464.mm.1 |
TC0600003038.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600023464.mm.1 |
NM_198884.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600023464.mm.1 |
NM_198884.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600023453.mm.1 |
TC0600003038.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600023453.mm.1 |
NM_198884.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600023453.mm.1 |
NM_198884.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600023461.mm.1 |
TC0600003038.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600023461.mm.1 |
NM_198884.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600023461.mm.1 |
NM_198884.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600023462.mm.1 |
TC0600003038.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600023462.mm.1 |
NM_198884.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600023462.mm.1 |
NM_198884.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600023456.mm.1 |
TC0600003038.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600023456.mm.1 |
NM_198884.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600023456.mm.1 |
NM_198884.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600023459.mm.1 |
TC0600003038.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600023459.mm.1 |
NM_198884.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600023459.mm.1 |
NM_198884.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600023463.mm.1 |
TC0600003038.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600023463.mm.1 |
NM_198884.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600023463.mm.1 |
NM_198884.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600023466.mm.1 |
TC0600003038.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600023466.mm.1 |
NM_198884.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600023466.mm.1 |
NM_198884.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600023449.mm.1 |
TC0600003038.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600023449.mm.1 |
NM_198884.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600023449.mm.1 |
NM_198884.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600023458.mm.1 |
TC0600003038.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600023458.mm.1 |
NM_198884.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600023458.mm.1 |
NM_198884.1 |
Refseq |