Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001160.3 | NP_001001160.1 | 9 - 10 | 2669 | 86 | |
NM_001289674.1 | NP_001276603.1 | 8 - 9 | 2500 | 86 | ||
NM_001289675.1 | NP_001276604.1 | 8 - 9 | 2494 | 86 | ||
GenBank mRNA | AK129466.1 | - | 7 - 8 | 2046 | 86 | |
AK147230.1 | - | 9 - 10 | 2561 | 86 | ||
AK147681.1 | - | 8 - 9 | 2359 | 86 | ||
BC061475.1 | - | 6 - 7 | 1141 | 86 | ||
BC070445.1 | - | 8 - 9 | 2500 | 30536738 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |