Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trmt11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028604.2 | NP_082880.2 | 3 - 4 | 295 | 145 | |
GenBank mRNA | AF532979.1 | - | 3 - 4 | 295 | 145 | |
AK010722.1 | - | 3 - 4 | 237 | 145 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |