Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cenpw | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109747.1 | NP_001103217.1 | 1 - 2 | 176 | 98 | |
GenBank mRNA | AK005988.1 | - | 1 - 2 | 158 | 98 | |
AK011700.1 | - | 1 - 2 | 158 | 98 | ||
AK141272.1 | - | 1 - 2 | 158 | 98 | ||
BC050071.1 | - | 128 | 98 | |||
BC147188.1 | - | 1 - 2 | 173 | 9056212 | 98 | |
BC147189.1 | - | 1 - 2 | 173 | 9056237 | 98 | |
BC147710.1 | - | 1 - 2 | 173 | 9087469 | 98 | |
BC147721.1 | - | 1 - 2 | 173 | 9087480 | 98 | |
BG804382.1 | - | 1 - 2 | 176 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Probe spans exons |