Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp6v1b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134157.2 | NP_598918.1 | 11 - 12 | 1212 | 66 | |
GenBank mRNA | AF435091.1 | - | 11 - 12 | 1212 | 66 | |
AK052604.1 | - | 11 - 12 | 1183 | 66 | ||
AK052707.1 | - | 11 - 12 | 1174 | 66 | ||
AK078810.1 | - | 11 - 12 | 1240 | 66 | ||
AK085549.1 | - | 11 - 12 | 1213 | 66 | ||
AK168980.1 | - | 11 - 12 | 1240 | 66 | ||
BC004789.1 | - | 8 - 9 | 806 | 66 | ||
BC017127.1 | - | 11 - 12 | 1197 | 4489144 | 66 | |
BC062202.1 | - | 11 - 12 | 1171 | 2650839 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel Transporter Mammalian Gene Collection Mus musculus Probe spans exons |