Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ghrl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001286405.1 | NP_001273334.1 | 1 - 2 | 130 | 69 | |
NM_021488.5 | NP_067463.2 | 2 - 3 | 178 | 69 | ||
GenBank mRNA | AB035701.1 | - | 1 - 2 | 120 | 69 | |
AK008658.1 | - | 2 - 3 | 148 | 69 | ||
AK008860.1 | - | 2 - 3 | 148 | 69 | ||
AY179430.1 | - | 1 - 2 | 120 | 69 | ||
BC094911.1 | - | 1 - 2 | 134 | 69 | ||
BC132230.1 | - | 1 - 2 | 144 | 40130507 | 69 | |
BC132232.1 | - | 1 - 2 | 144 | 40130509 | 69 | |
FJ914266.1 | - | 1 - 2 | 184 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |