Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdk19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198164.3 | NP_937807.2 | 5 - 6 | 797 | 77 | |
GenBank mRNA | AK053651.1 | - | 5 - 6 | 770 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mus musculus Probe spans exons |