Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Themis | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AB509340.1 | - | 3 - 4 | 799 | 120 | |
AK040594.1 | - | 3 - 4 | 978 | 120 | ||
AK041386.1 | - | 3 - 4 | 1015 | 120 | ||
AK041562.1 | - | 3 - 4 | 904 | 120 | ||
AK088136.1 | - | 3 - 4 | 980 | 120 | ||
AK138701.1 | - | 3 - 4 | 978 | 120 | ||
AK169558.1 | - | 4 - 5 | 910 | 120 | ||
BC125486.1 | - | 3 - 4 | 861 | 40130001 | 120 | |
BC125488.1 | - | 3 - 4 | 861 | 40130003 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Mus musculus Probe spans exons |