Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gdi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010273.4 | NP_034403.1 | 7 - 8 | 1201 | 61 | |
GenBank mRNA | AF251042.1 | - | 7 - 8 | 870 | 61 | |
AK028880.1 | - | 7 - 8 | 958 | 61 | ||
AK032869.1 | - | 6 - 7 | 1207 | 61 | ||
AK156224.1 | - | 7 - 8 | 969 | 61 | ||
AK171216.1 | - | 7 - 8 | 958 | 61 | ||
AK216611.1 | - | 2 - 3 | 165 | 61 | ||
BC013758.1 | - | 7 - 8 | 882 | 4501084 | 61 | |
BC025606.1 | - | 7 - 8 | 968 | 61 | ||
BC037598.1 | - | 7 - 8 | 935 | 5249271 | 61 | |
U07950.1 | - | 3 - 4 | 444 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |