Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hsf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008297.3 | NP_032323.3 | 6 - 7 | 718 | 130 | |
GenBank mRNA | AK052270.1 | - | 6 - 7 | 617 | 130 | |
AK053976.1 | - | 6 - 7 | 719 | 130 | ||
BC018414.1 | - | 6 - 7 | 640 | 3990765 | 130 | |
X61754.1 | - | 6 - 7 | 752 | 130 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |