Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4831410D14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK029182.1 | - | 2 - 3 | 399 | 64 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | RAN binding protein 17 | Ranbp17 | Anti-sense | Chr. 11 - 33211794 - 33514750 on NCBI Build GRCm38 | ||
Downstream |
acidic (leucine-rich) nuclear phosphoprotein 32 family, pseudogene |
Anp32-ps |
Anp32c, Anp32c-ps, Anp32cl, EG448829 |
176.3kb | Anti-sense | Chr. 11 - 33394905 - 33395397 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length less than or equal to 70 Mus musculus Probe spans exons |