Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bclaf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025392.1 | NP_001020563.1 | 3 - 4 | 357 | 142 | |
NM_001025393.1 | NP_001020564.1 | 3 - 4 | 357 | 142 | ||
GenBank mRNA | AK011802.1 | - | 3 - 4 | 297 | 142 | |
AK149724.1 | - | 3 - 4 | 310 | 142 | ||
AK150017.1 | - | 3 - 4 | 310 | 142 | ||
AK161692.1 | - | 3 - 4 | 355 | 142 | ||
AK162896.1 | - | 3 - 4 | 315 | 142 | ||
BC028963.1 | - | 3 - 4 | 289 | 142 | ||
BC029682.1 | - | 3 - 4 | 288 | 142 | ||
BC086624.1 | - | 3 - 4 | 342 | 142 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |