Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bclaf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025392.1 | NP_001020563.1 | 6 - 7 | 2096 | 105 | |
NM_001025393.1 | NP_001020564.1 | 6 - 7 | 2096 | 105 | ||
NM_153787.2 | NP_722482.1 | 6 - 7 | 2090 | 105 | ||
GenBank mRNA | AK088741.1 | - | 6 - 7 | 2031 | 105 | |
AK129071.1 | - | 3 - 4 | 1380 | 105 | ||
AK141834.1 | - | 6 - 7 | 2043 | 105 | ||
AK149724.1 | - | 6 - 7 | 2049 | 105 | ||
AK150017.1 | - | 6 - 7 | 2049 | 105 | ||
AK161608.1 | - | 985 | 105 | |||
AK161692.1 | - | 6 - 7 | 2094 | 105 | ||
BC029682.1 | - | 6 - 7 | 2027 | 105 | ||
BC034300.1 | - | 6 - 7 | 2022 | 4507876 | 105 | |
BC086624.1 | - | 6 - 7 | 2078 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |