Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023816.2 | NP_076305.2 | 1 - 2 | 445 | 63 | |
GenBank mRNA | AF294328.1 | - | 1 - 2 | 447 | 63 | |
AK005925.1 | - | 1 - 2 | 419 | 63 | ||
AK132751.1 | - | 1 - 2 | 446 | 63 | ||
BC120815.1 | - | 1 - 2 | 223 | 63 | ||
BC120817.1 | - | 1 - 2 | 220 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |