Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plxna3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008883.2 | NP_032909.2 | 2 - 3 | 773 | 81 | |
GenBank mRNA | BC093482.1 | - | 2 - 3 | 756 | 30945372 | 81 |
D86950.1 | - | 2 - 3 | 739 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |