Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sytl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031393.2 | NP_113570.2 | 7 - 8 | 785 | 93 | |
GenBank mRNA | AB050741.1 | - | 7 - 8 | 764 | 93 | |
AK078868.1 | - | 7 - 8 | 774 | 93 | ||
AK131162.1 | - | 7 - 8 | 742 | 93 | ||
BC005623.1 | - | 7 - 8 | 785 | 3155535 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |