Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400030407.mm.1 |
TC0400003711.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400030407.mm.1 |
NM_001145955.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400030407.mm.1 |
NM_001145955.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400030407.mm.1 |
NM_178698.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400030407.mm.1 |
NM_178698.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400030406.mm.1 |
TC0400003711.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400030406.mm.1 |
NM_001145955.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400030406.mm.1 |
NM_001145955.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400030406.mm.1 |
NM_001145956.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400030406.mm.1 |
NM_001145956.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400030406.mm.1 |
NM_178698.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400030406.mm.1 |
NM_178698.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400030404.mm.1 |
TC0400003711.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400030404.mm.1 |
NM_001145955.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400030404.mm.1 |
NM_001145955.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400030404.mm.1 |
NM_001145956.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400030404.mm.1 |
NM_001145956.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400030404.mm.1 |
NM_178698.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400030404.mm.1 |
NM_178698.5 |
Refseq |