Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps6ka1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285505.1 | NP_001272434.1 | 18 - 19 | 1886 | 74 | |
NM_001285506.1 | NP_001272435.1 | 17 - 18 | 1770 | 74 | ||
NM_009097.5 | NP_033123.2 | 18 - 19 | 1919 | 74 | ||
GenBank mRNA | AK132856.1 | - | 16 - 17 | 1956 | 74 | |
AK148208.1 | - | 18 - 19 | 1912 | 74 | ||
AK160571.1 | - | 6 - 7 | 714 | 74 | ||
AK167790.1 | - | 17 - 18 | 1757 | 74 | ||
BC049076.1 | - | 18 - 19 | 1870 | 74 | ||
BC094470.1 | - | 18 - 19 | 1872 | 5098650 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |