Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK052762.1 | - | 1 - 2 | 67 | 118 | |
AK085717.1 | - | 1 - 2 | 157 | 118 | ||
AK154613.1 | - | 1 - 2 | 213 | 118 | ||
AK163171.1 | - | 1 - 2 | 159 | 118 | ||
AK170641.1 | - | 1 - 2 | 199 | 118 | ||
BB619031.1 | - | 1 - 2 | 260 | 118 | ||
BC022976.1 | - | 1 - 2 | 145 | 5362925 | 118 | |
BC024940.1 | - | 1 - 2 | 143 | 118 | ||
BC071258.1 | - | 1 - 2 | 78 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |