Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trit1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025873.2 | NP_080149.2 | 4 - 5 | 574 | 94 | |
GenBank mRNA | AK003556.1 | - | 4 - 5 | 575 | 94 | |
AK010176.1 | - | 4 - 5 | 574 | 94 | ||
AK128921.1 | - | 4 - 5 | 573 | 94 | ||
BC019812.1 | - | 4 - 5 | 543 | 5042856 | 94 | |
BC051040.1 | - | 4 - 5 | 550 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |