Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nt5c1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085502.1 | NP_001078971.1 | 1 - 2 | 135 | 60 | |
GenBank mRNA | BC147308.1 | - | 1 - 2 | 184 | 9056382 | 60 |
BC147309.1 | - | 1 - 2 | 184 | 9056378 | 60 | |
BC147730.1 | - | 1 - 2 | 179 | 9056679 | 60 | |
BC147733.1 | - | 1 - 2 | 179 | 9056682 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |