Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr92 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178909.4 | NP_849240.1 | 1 - 2 | 251 | 81 | |
GenBank mRNA | AK033294.1 | - | 1 - 2 | 197 | 81 | |
AK047691.1 | - | 1 - 2 | 226 | 81 | ||
AK088218.1 | - | 1 - 2 | 226 | 81 | ||
AK142228.1 | - | 1 - 2 | 197 | 81 | ||
BC096540.1 | - | 1 - 2 | 180 | 6403681 | 81 | |
BY794211.1 | - | 1 - 2 | 252 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |