Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ifngr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010511.2 | NP_034641.1 | 4 - 5 | 700 | 97 | |
GenBank mRNA | AF128216.1 | - | 4 - 5 | 651 | 97 | |
AF128217.1 | - | 4 - 5 | 651 | 97 | ||
AK087525.1 | - | 4 - 5 | 689 | 97 | ||
AK150503.1 | - | 4 - 5 | 649 | 97 | ||
AK151079.1 | - | 4 - 5 | 654 | 97 | ||
AK152085.1 | - | 4 - 5 | 649 | 97 | ||
AK152384.1 | - | 4 - 5 | 649 | 97 | ||
AK153084.1 | - | 4 - 5 | 649 | 97 | ||
AK153347.1 | - | 4 - 5 | 652 | 97 | ||
AK159359.1 | - | 4 - 5 | 654 | 97 | ||
AK169593.1 | - | 4 - 5 | 669 | 97 | ||
AY157836.1 | - | 4 - 5 | 651 | 97 | ||
AY157837.1 | - | 4 - 5 | 651 | 97 | ||
BC132599.1 | - | 4 - 5 | 601 | 40130876 | 97 | |
BC138617.1 | - | 4 - 5 | 601 | 8861637 | 97 | |
J05265.1 | - | 4 - 5 | 677 | 97 | ||
M25764.1 | - | 4 - 5 | 602 | 97 | ||
M26711.1 | - | 4 - 5 | 677 | 97 | ||
M28233.1 | - | 4 - 5 | 673 | 97 | ||
M28995.1 | - | 4 - 5 | 578 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |