Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1000002024.mm.1 |
NM_001081208.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000002024.mm.1 |
NM_001081208.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000002024.mm.1 |
NM_001253355.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000002024.mm.1 |
NM_001253355.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000002024.mm.1 |
NM_001253356.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000002024.mm.1 |
NM_001253356.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000002024.mm.1 |
TC1000000306.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000002027.mm.1 |
NM_001081208.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000002027.mm.1 |
NM_001081208.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000002027.mm.1 |
NM_001253355.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000002027.mm.1 |
NM_001253355.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000002027.mm.1 |
NM_001253356.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000002027.mm.1 |
NM_001253356.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000002027.mm.1 |
TC1000000306.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000002026.mm.1 |
NM_001081208.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000002026.mm.1 |
NM_001081208.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000002026.mm.1 |
NM_001253355.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000002026.mm.1 |
NM_001253355.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000002026.mm.1 |
NM_001253356.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000002026.mm.1 |
NM_001253356.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000002026.mm.1 |
TC1000000306.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000002028.mm.1 |
NM_001253355.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000002028.mm.1 |
NM_001253355.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000002028.mm.1 |
TC1000000306.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000002025.mm.1 |
NM_001081208.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000002025.mm.1 |
NM_001081208.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000002025.mm.1 |
NM_001253355.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000002025.mm.1 |
NM_001253355.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000002025.mm.1 |
NM_001253356.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000002025.mm.1 |
NM_001253356.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000002025.mm.1 |
TC1000000306.mm.1 |
MTA TC |