Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gabrp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146017.3 | NP_666129.1 | 8 - 9 | 1035 | 76 | |
GenBank mRNA | AK028946.1 | - | 8 - 9 | 1035 | 76 | |
AK160577.1 | - | 8 - 9 | 1039 | 76 | ||
BC023693.1 | - | 8 - 9 | 1022 | 5344479 | 76 | |
BC025550.1 | - | 8 - 9 | 980 | 5320737 | 76 | |
BC027245.1 | - | 8 - 9 | 998 | 3602400 | 76 | |
BC027839.1 | - | 4 - 5 | 587 | 76 | ||
BC031196.1 | - | 8 - 9 | 1009 | 5059564 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Ion Channel Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |