Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Txlnb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138628.3 | NP_619534.2 | 5 - 6 | 1107 | 76 | |
GenBank mRNA | AF422244.1 | - | 5 - 6 | 1044 | 76 | |
AF422245.1 | - | 5 - 6 | 1044 | 76 | ||
AK084639.1 | - | 5 - 6 | 1108 | 76 | ||
AK085904.1 | - | 5 - 6 | 1003 | 76 | ||
AK137442.1 | - | 5 - 6 | 1022 | 76 | ||
BC141023.1 | - | 5 - 6 | 1020 | 9055635 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |