Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nprl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001284359.1 | NP_001271288.1 | 8 - 9 | 976 | 76 | |
NM_001284360.1 | NP_001271289.1 | 6 - 7 | 807 | 76 | ||
NM_181569.3 | NP_853547.1 | 7 - 8 | 937 | 76 | ||
NR_104306.1 | 7 - 8 | 855 | 76 | |||
GenBank mRNA | AK036405.1 | - | 7 - 8 | 846 | 76 | |
AK036633.1 | - | 6 - 7 | 906 | 76 | ||
AK039600.1 | - | 6 - 7 | 783 | 76 | ||
AK047190.1 | - | 8 - 9 | 885 | 76 | ||
AK081902.1 | - | 7 - 8 | 848 | 76 | ||
AK086378.1 | - | 750 | 76 | |||
AK090290.1 | - | 715 | 76 | |||
AK154884.1 | - | 7 - 8 | 807 | 76 | ||
BC064471.1 | - | 7 - 8 | 937 | 6334386 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |