Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nprl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001284359.1 | NP_001271288.1 | 9 - 10 | 71 | ||
NM_001284360.1 | NP_001271289.1 | 7 - 8 | 71 | |||
NM_181569.3 | NP_853547.1 | 8 - 9 | 71 | |||
GenBank mRNA | AK036405.1 | - | 8 - 9 | 71 | ||
AK036633.1 | - | 7 - 8 | 71 | |||
AK039600.1 | - | 7 - 8 | 71 | |||
AK047190.1 | - | 9 - 10 | 71 | |||
AK081902.1 | - | 8 - 9 | 71 | |||
AK086378.1 | - | 71 | ||||
AK090290.1 | - | 71 | ||||
BC064471.1 | - | 8 - 9 | 6334386 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |