Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc6a8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142809.1 | NP_001136281.1 | 4 - 5 | 1432 | 91 | |
NM_001142810.1 | NP_001136282.1 | 4 - 5 | 1432 | 91 | ||
NM_133987.2 | NP_598748.1 | 4 - 5 | 1432 | 91 | ||
GenBank mRNA | AB077327.1 | - | 4 - 5 | 780 | 91 | |
AF459435.1 | - | 4 - 5 | 1415 | 91 | ||
BC024444.1 | - | 3 - 4 | 445 | 91 | ||
BC029000.1 | - | 2 - 3 | 195 | 91 | ||
BC049801.1 | - | 4 - 5 | 607 | 91 | ||
BC141067.1 | - | 5 - 6 | 1173 | 9055679 | 91 | |
BC145319.1 | - | 5 - 6 | 837 | 9053874 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |