Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lmtk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005511.3 | NP_001005511.2 | 5 - 6 | 512 | 56 | |
NM_001290990.1 | NP_001277919.1 | 4 - 5 | 667 | 56 | ||
NR_110987.1 | 5 - 6 | 512 | 56 | |||
GenBank mRNA | AB288873.1 | - | 5 - 6 | 506 | 56 | |
BC057032.1 | - | 4 - 5 | 400 | 56 | ||
BC059845.1 | - | 5 - 6 | 721 | 56 | ||
BC083185.1 | - | 4 - 5 | 667 | 6855048 | 56 | |
BC094377.1 | - | 5 - 6 | 512 | 30536822 | 56 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |