Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sgsm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001162965.1 | NP_001156437.1 | 8 - 9 | 896 | 65 | |
NM_001254731.1 | NP_001241660.1 | 8 - 9 | 896 | 65 | ||
NM_172718.3 | NP_766306.2 | 8 - 9 | 896 | 65 | ||
GenBank mRNA | AK044716.1 | - | 8 - 9 | 850 | 65 | |
AK053555.1 | - | 8 - 9 | 830 | 65 | ||
AK170021.1 | - | 8 - 9 | 888 | 65 | ||
BC138049.1 | - | 8 - 9 | 822 | 8861068 | 65 | |
BC138050.1 | - | 8 - 9 | 822 | 8861069 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons |