Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028627.2 | NP_082903.2 | 14 - 15 | 2609 | 75 | |
GenBank mRNA | AK158685.1 | - | 4 - 5 | 415 | 75 | |
AK158851.1 | - | 4 - 5 | 541 | 75 | ||
AK220438.1 | - | 9 - 10 | 2726 | 75 | ||
BC043135.1 | - | 10 - 11 | 1438 | 75 | ||
BC057353.1 | - | 14 - 15 | 2610 | 75 | ||
BC138652.1 | - | 12 - 13 | 2468 | 8861672 | 75 | |
BC145352.1 | - | 12 - 13 | 2471 | 9053908 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |